Merck
CCNE1 amplification is synthetic lethal with PKMYT1 kinase inhibition.
Nature (2022-04-22)
David Gallo, Jordan T F Young, Jimmy Fourtounis, Giovanni Martino, Alejandro Álvarez-Quilón, Cynthia Bernier, Nicole M Duffy, Robert Papp, Anne Roulston, Rino Stocco, Janek Szychowski, Artur Veloso, Hunain Alam, Prasamit S Baruah, Alexanne Bonneau Fortin, Julian Bowlan, Natasha Chaudhary, Jessica Desjardins, Evelyne Dietrich, Sara Fournier, Chloe Fugère-Desjardins, Theo Goullet de Rugy, Marie-Eve Leclaire, Bingcan Liu, Vivek Bhaskaran, Yael Mamane, Henrique Melo, Olivier Nicolas, Akul Singhania, Rachel K Szilard, Ján Tkáč, Shou Yun Yin, Stephen J Morris, Michael Zinda, C Gary Marshall, Daniel Durocher
ABSTRACT
Amplification of the CCNE1 locus on chromosome 19q12 is prevalent in multiple tumour types, particularly in high-grade serous ovarian cancer, uterine tumours and gastro-oesophageal cancers, where high cyclin E levels are associated with genome instability, whole-genome doubling and resistance to cytotoxic and targeted therapies1-4. To uncover therapeutic targets for tumours with CCNE1 amplification, we undertook genome-scale CRISPR-Cas9-based synthetic lethality screens in cellular models of CCNE1 amplification. Here we report that increasing CCNE1 dosage engenders a vulnerability to the inhibition of the PKMYT1 kinase, a negative regulator of CDK1. To inhibit PKMYT1, we developed RP-6306, an orally bioavailable and selective inhibitor that shows single-agent activity and durable tumour regressions when combined with gemcitabine in models of CCNE1 amplification. RP-6306 treatment causes unscheduled activation of CDK1 selectively in CCNE1-overexpressing cells, promoting early mitosis in cells undergoing DNA synthesis. CCNE1 overexpression disrupts CDK1 homeostasis at least in part through an early activation of the MMB-FOXM1 mitotic transcriptional program. We conclude that PKMYT1 inhibition is a promising therapeutic strategy for CCNE1-amplified cancers.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Anti-Cdk2 Antibody, clone AN4.3, clone AN4.3, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-B-Myb Antibody, clone LX015.1, clone LX015.1, from mouse
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Histone H1 human
Sigma-Aldrich
Goat Serum Donor Herd, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-α-Actinin Antibody, clone AT6/172, clone AT6/172, Upstate®, from mouse