Merck
  • Oligodendrocyte precursor cell specification is regulated by bidirectional neural progenitor-endothelial cell crosstalk.
Oligodendrocyte precursor cell specification is regulated by bidirectional neural progenitor-endothelial cell crosstalk.
Nature neuroscience (2021-01-30)
Isidora Paredes, José Ricardo Vieira, Bhavin Shah, Carla F Ramunno, Julia Dyckow, Heike Adler, Melanie Richter, Geza Schermann, Evangelia Giannakouri, Lucas Schirmer, Hellmut G Augustin, Carmen Ruiz de Almodóvar
ABSTRACT
Neural-derived signals are crucial regulators of CNS vascularization. However, whether the vasculature responds to these signals by means of elongating and branching or in addition by building a feedback response to modulate neurodevelopmental processes remains unknown. In this study, we identified bidirectional crosstalk between the neural and the vascular compartment of the developing CNS required for oligodendrocyte precursor cell specification. Mechanistically, we show that neural progenitor cells (NPCs) express angiopoietin-1 (Ang1) and that this expression is regulated by Sonic hedgehog. We demonstrate that NPC-derived Ang1 signals to its receptor, Tie2, on endothelial cells to induce the production of transforming growth factor beta 1 (TGFβ1). Endothelial-derived TGFβ1, in turn, acts as an angiocrine molecule and signals back to NPCs to induce their commitment toward oligodendrocyte precursor cells. This work demonstrates a true bidirectional collaboration between NPCs and the vasculature as a critical regulator of oligodendrogenesis.
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
SANT-1, A potent, cell-permeable antagonist of the Shh signaling pathway by binding directly to Smoothened, a distant relative of G protein-coupled receptors.