Merck
  • N1-methyladenosine methylation in tRNA drives liver tumourigenesis by regulating cholesterol metabolism.
N1-methyladenosine methylation in tRNA drives liver tumourigenesis by regulating cholesterol metabolism.
Nature communications (2021-11-04)
Yanying Wang, Jing Wang, Xiaoyu Li, Xushen Xiong, Jianyi Wang, Ziheng Zhou, Xiaoxiao Zhu, Yang Gu, Dan Dominissini, Lei He, Yong Tian, Chengqi Yi, Zusen Fan
ABSTRACT
Hepatocellular carcinoma (HCC) accounts for the majority of primary liver cancers and is characterized by high recurrence and heterogeneity, yet its mechanism is not well understood. Here we show that N1-methyladenosine methylation (m1A) in tRNA is remarkably elevated in hepatocellular carcinoma (HCC) patient tumour tissues. Moreover, m1A methylation signals are increased in liver cancer stem cells (CSCs) and are negatively correlated with HCC patient survival. TRMT6 and TRMT61A, forming m1A methyltransferase complex, are highly expressed in advanced HCC tumours and are negatively correlated with HCC survival. TRMT6/TRMT61A-mediated m1A methylation is required for liver tumourigenesis. Mechanistically, TRMT6/TRMT61A elevates the m1A methylation in a subset of tRNA to increase PPARδ translation, which in turn triggers cholesterol synthesis to activate Hedgehog signaling, eventually driving self-renewal of liver CSCs and tumourigenesis. Finally, we identify a potent inhibitor against TRMT6/TRMT61A complex that exerts effective therapeutic effect on liver cancer.
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
PPARβ/δ Antagonist, GSK3787, The PPARβ/δAntagonist II, PT-S58, also referenced under CAS 188591-46-0, controls the biological activity of PPARβ/δ. This small molecule/inhibitor is primarily used for Biochemicals applications.
Pricing and availability is not currently available.
Benserazide hydrochloride, British Pharmacopoeia (BP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
GW501516, ≥98% (HPLC)
Pricing and availability is not currently available.