Merck
  • Codonopsis pilosula Polysaccharides Alleviate Aβ 1-40-Induced PC12 Cells Energy Dysmetabolism via CD38/NAD+ Signaling Pathway.
Codonopsis pilosula Polysaccharides Alleviate Aβ 1-40-Induced PC12 Cells Energy Dysmetabolism via CD38/NAD+ Signaling Pathway.
Current Alzheimer research (2021-06-10)
Yi R Hu, San L Xing, Chuan Chen, Ding Z Shen, Jiu L Chen
ABSTRACT
Alzheimer's disease (AD) is the most common type of dementia and has a complex pathogenesis with no effective treatment. Energy metabolism disorders, as an early pathological event of AD,have attracted attention as a promising area of AD research. Codonopsis pilosula Polysaccharides are the main effective components of Codonopsis pilosula, which have been demonstrated to regulate energy metabolism. In order to further study the roles and mechanisms of Codonopsis pilosula polysaccharides in AD, this study used an Aβ1-40-induced PC12 cells model to study the protective effects of Codonopsis pilosula polysaccharides and their potential mechanisms in improving energy metabolism dysfunction. The results showed that Aβ1-40 induced a decrease in PC12 cells viability, energy metabolism molecules (ATP, NAD+, and NAD+/NADH) and Mitochondrial Membrane Potential (MMP) and an increase in ROS. Additionally, it was found that Aβ1-40 increased CD38 expression related to NAD+ homeostasis, whereas Silent Information Regulation 2 homolog1 (SIRT1, SIRT3), Peroxisome proliferator-activated receptor γ coactivator 1-α (PGC-1α) and SIRT3 activity were decreased. Codonopsis pilosula polysaccharides increased NAD+, NAD+/NADH, SIRT3, SIRT1, and PGC-1α related to NAD+, thus partially recovering ATP. Our findings reveal that Codonopsis pilosula polysaccharides protected PC12 cells from Aβ1-40-induced damage, suggesting that these components of the Codonopsis pilosula herb may represent an early treatment option for AD patients.
Sigma-Aldrich
Amyloid β Protein Fragment 1-40, ≥90% (HPLC), powder