Merck
  • Molecular analysis of antimicrobial resistance mechanisms in Neisseria gonorrhoeae isolates from Ontario, Canada.
Molecular analysis of antimicrobial resistance mechanisms in Neisseria gonorrhoeae isolates from Ontario, Canada.
Antimicrobial agents and chemotherapy (2010-11-26)
Vanessa G Allen, David J Farrell, Anuradha Rebbapragada, Jingyuan Tan, Nathalie Tijet, Stephen J Perusini, Lynn Towns, Stephen Lo, Donald E Low, Roberto G Melano
ABSTRACT
Surveillance of gonococcal antimicrobial resistance and the molecular characterization of the mechanisms underlying these resistance phenotypes are essential in order to establish correct empirical therapies, as well as to describe the emergence of new mechanisms in local bacterial populations. To address these goals, 149 isolates were collected over a 1-month period (October-November 2008) at the Ontario Public Health Laboratory, Toronto, Canada, and susceptibility profiles (8 antibiotics) were examined. Mutations in previously identified targets or the presence of some enzymes related to resistance (r), nonsusceptibility (ns) (resistant plus intermediate categories), or reduced susceptibility (rs) to the antibiotics tested were also studied. A significant proportion of nonsusceptibility to penicillin (PEN) (89.2%), tetracycline (TET) (72.3%), ciprofloxacin (CIP) (29%), and macrolides (erythromycin [ERY] and azithromycin; 22.3%) was found in these strains. Multidrug resistance was observed in 18.8% of the collection. Although all the strains were susceptible to spectinomycin and extended-spectrum cephalosporins (ESC) (ceftriaxone and cefixime), 9.4% of them displayed reduced susceptibility to extended-spectrum cephalosporins. PBP 2 mosaic structures were found in all of these ESC(rs) isolates. Alterations in the mtrR promoter, MtrR repressor (TET(r), PEN(ns), ESC(rs), and ERY(ns)), porin PIB (TET(r) and PEN(ns)), and ribosomal protein S10 (TET(r)) and double mutations in gyrA and parC quinolone resistance-determining regions (QRDRs) (CIP(r)) were associated with and presumably responsible for the resistance phenotypes observed. This is the first description of ESC(rs) in Canada. The detection of this phenotype indicates a change in the epidemiology of this resistance and highlights the importance of continued surveillance to preserve the last antimicrobial options available.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg