Merck
  • Novel natural product-based cinnamates and their thio and thiono analogs as potent inhibitors of cell adhesion molecules on human endothelial cells.
Novel natural product-based cinnamates and their thio and thiono analogs as potent inhibitors of cell adhesion molecules on human endothelial cells.
European journal of medicinal chemistry (2011-10-01)
Sarvesh Kumar, Brajendra K Singh, Pragya Arya, Shashwat Malhotra, Rajesh Thimmulappa, Ashok K Prasad, Erik Van der Eycken, Carl E Olsen, Anthony L DePass, Shyam Biswal, Virinder S Parmar, Balaram Ghosh
ABSTRACT
In the present study, we report the design and synthesis of novel analogs of cinnamates, thiocinnamates and thionocinnamates and evaluated the potencies of these analogs to inhibit TNF-α induced ICAM-1 expression on human endothelial cells. By using whole cell-ELISA, our screening data demonstrated that ethyl 3',4',5'-trimethoxythionocinnamate (ETMTC) is the most potent inhibitor of TNF-α induced ICAM-1, VCAM-1 and E-selectin. As functional consequences, ETMTC abrogated TNF-α induced adhesion of neutrophils to the endothelial monolayer. Structure-activity relationship studies revealed the critical role of the chain-length of the alkyl group in the alcohol moiety, number of methoxy groups in the aromatic ring of the cinnamoyl moiety and the presence of the α, β- C-C double bond in the thiocinnamates and thionocinnamates.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
Ethyl cinnamate, natural, ≥95%, FG
Sigma-Aldrich
Ethyl cinnamate, 99%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Ethyl cinnamate, ≥98%, stabilized, FCC, FG
Sigma-Aldrich
Benzyl cinnamate, 99%
Sigma-Aldrich
Benzyl cinnamate, ≥98%, stabilized
Sigma-Aldrich
5-Aminosalicylic acid, 95%