Merck
  • HIV-infected macrophages resist efficient NK cell-mediated killing while preserving inflammatory cytokine responses.
HIV-infected macrophages resist efficient NK cell-mediated killing while preserving inflammatory cytokine responses.
Cell host & microbe (2021-02-12)
Kiera L Clayton, Geetha Mylvaganam, Alonso Villasmil-Ocando, Heather Stuart, Marcela V Maus, Mohammad Rashidian, Hidde L Ploegh, Bruce D Walker
ABSTRACT
Natural killer (NK) cells are innate cytolytic effectors that target HIV-infected CD4+ T cells. In conjunction with antibodies recognizing the HIV envelope, NK cells also eliminate HIV-infected targets through antibody-dependent cellular cytotoxicity (ADCC). However, how these NK cell functions impact infected macrophages is less understood. We show that HIV-infected macrophages resist NK cell-mediated killing. Compared with HIV-infected CD4+ T cells, initial innate NK cell interactions with HIV-infected macrophages skew the response toward cytokine production, rather than release of cytolytic contents, causing inefficient elimination of infected macrophages. Studies with chimeric antigen receptor (CAR) T cells demonstrate that the viral envelope is equally accessible on CD4+ T cells and macrophages. Nonetheless, ADCC against macrophages is muted compared with ADCC against CD4+ T cells. Thus, HIV-infected macrophages employ mechanisms to evade immediate cytolytic NK cell function while preserving inflammatory cytokine responses. These findings emphasize the importance of eliminating infected macrophages for HIV cure efforts.
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-M, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Corning® Costar® Ultra-Low Attachment Multiple Well Plate, size 6 well, flat bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid
Corning® Costar® Ultra-Low Attachment Multiple Well Plate, size 24 well, flat bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Anti-Siglec-1 (CD169) Antibody, clone 5F1.1, clone 5F1.1, from mouse