Merck
  • N-Glycosylation at Asn695 might suppress inducible nitric oxide synthase activity by disturbing electron transfer.
N-Glycosylation at Asn695 might suppress inducible nitric oxide synthase activity by disturbing electron transfer.
Acta biochimica et biophysica Sinica (2020-11-25)
Jianghong Yan, Fei-Fei Shang, An He, Shupeng Hu, Suxin Luo, Yong Xia
ABSTRACT
Inducible nitric oxide synthase (iNOS) plays critical roles in the inflammatory response and host defense. Previous research on iNOS regulation mainly focused on its gene expression level, and much less is known about the regulation of iNOS function by N-glycosylation. In this study, we report for the first time that iNOS is N-glycosylated in vitro and in vivo. Mass spectrometry studies identified Asn695 as an N-glycosylation site of murine iNOS. Mutating Asn695 to Gln695 yields an iNOS that exhibits greater enzyme activity. The essence of nitric oxide synthase catalytic reaction is electron transfer process, which involves a series of conformational changes, and the linker between the flavin mononucleotide-binding domain and the flavin adenine dinucleotide-binding domain plays vital roles in the conformational changes. Asn695 is part of the linker, so we speculated that attachment of N-glycan to the Asn695 residue might inhibit activity by disturbing electron transfer. Indeed, our NADPH consumption results demonstrated that N-glycosylated iNOS consumes NADPH more slowly. Taken together, our results indicate that iNOS is N-glycosylated at its Asn695 residue and N-glycosylation of Asn695 might suppress iNOS activity by disturbing electron transfer.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Supelco
Dowex 50W X8, hydrogen form, strongly acidic, 200-400 mesh
Pricing and availability is not currently available.
Sigma-Aldrich
Anthranilamide, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Lectin from Triticum vulgaris (wheat), lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), peroxidase conjugate, lyophilized powder
Pricing and availability is not currently available.