Merck
  • Lead identification of conformationally restricted β-lactam type combretastatin analogues: synthesis, antiproliferative activity and tubulin targeting effects.
Lead identification of conformationally restricted β-lactam type combretastatin analogues: synthesis, antiproliferative activity and tubulin targeting effects.
European journal of medicinal chemistry (2010-10-12)
Miriam Carr, Lisa M Greene, Andrew J S Knox, David G Lloyd, Daniela M Zisterer, Mary J Meegan
ABSTRACT
The synthesis and study of the structure-activity relationships of a series of rigid analogues of combretastatin A-4 are described which contain the 1,4-diaryl-2-azetidinone (β-lactam) ring system in place of the usual ethylene bridge present in the natural combretastatin stilbene products. The 1,4-diaryl-2-azetidinones are unsubstituted at C-3, or contain methyl substituent(s) at C-3. The most potent compounds 12d and 12e display antiproliferative activity at nanomolar concentrations when evaluated against the MCF-7 and MDA-MB-231 human breast carcinoma cell lines. 12d exerts antimitotic effects through an inhibition of tubulin polymerisation and subsequent G2/M arrest of the cell cycle in human MDA-MB-231 breast cancer cells, with similar activity to that of CA-4. These novel β-lactam compounds are identified as potentially useful scaffolds for the further development of antitumour agents which target tubulin.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, reagent grade
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC