Merck
  • Injectable thermosensitive chitosan-based hydrogel containing ferulic acid for treating peripheral arterial disease.
Injectable thermosensitive chitosan-based hydrogel containing ferulic acid for treating peripheral arterial disease.
Journal of tissue engineering and regenerative medicine (2020-08-09)
Chien-Ying Wang, Chen-Yuan Hsiao, Kun-Ling Tsai, Yung-Hsin Cheng
ABSTRACT
Peripheral arterial disease (PAD) affects more than 200 million people worldwide. Recent studies suggest that oxidative stress-related inflammation can lead to the initiation and progression of PAD. Ferulic acid (FA) is a natural phenolic compound and has been proven to have antioxidant and angiogenesis effects. In this study, thermosensitive chitosan-gelatin-based hydrogel was used as a delivery vehicle of FA. The effects of hydrogel encapsulating FA (FA-gel) have been demonstrated in vitro and in vivo. The results revealed that the developed hydrogel with porous structure could provide a sustained release of FA. Post-treatment of FA-gel effectively decreased the oxidative stress-induced damage in human umbilical vein endothelial cells via decreasing endogenous reactive oxygen species production, inflammation-related gene expression and apoptosis level. In the mouse hindlimb ischemia model, the results revealed that FA-gel could improve blood flow, muscle regeneration and decreases inflammation in veins. These results suggested that FA-gel may have a therapeutic potential in PAD.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A