Merck
  • Carbonic anhydrase activators: Activation of the beta-carbonic anhydrase from the pathogenic yeast Candida glabrata with amines and amino acids.
Carbonic anhydrase activators: Activation of the beta-carbonic anhydrase from the pathogenic yeast Candida glabrata with amines and amino acids.
Bioorganic & medicinal chemistry letters (2010-02-05)
Alessio Innocenti, Worraanong Leewattanapasuk, Gheorghe Manole, Andrea Scozzafava, Fritz A Mühlschlegel, Claudiu T Supuran
ABSTRACT
The protein encoded by the NCE103 gene of Candida glabrata, a beta-carbonic anhydrase (CA, EC 4.2.1.1) designated as CgCA, was investigated for its activation with amines and amino acids. CgCA was weakly activated by amino acids such as l-/d-His, l-Phe, l-DOPA, and l-Trp and by histamine or dopamine (K(A)s of 21.2-37microM) but more effectively activated by d-Phe, d-DOPA, d-Trp as well as serotonin, pyridyl-alkylamines, aminoethyl-piperazine/morpholine (K(A)s of 10.1-16.7microM). The best activators were l-/d-Tyr, with activation constants of 7.1-9.5microM. This study may bring a better understanding of the catalytic/activation mechanisms of beta-CAs from pathogenic fungi.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Tyrosine
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
SAFC
L-Histidine
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Tyrosine, ReagentPlus®, 99%