Merck
  • Propionate Enhances Cell Speed and Persistence to Promote Intestinal Epithelial Turnover and Repair.
Propionate Enhances Cell Speed and Persistence to Promote Intestinal Epithelial Turnover and Repair.
Cellular and molecular gastroenterology and hepatology (2020-11-26)
Anthony J Bilotta, Chunyan Ma, Wenjing Yang, Yanbo Yu, Yu Yu, Xiaojing Zhao, Zheng Zhou, Suxia Yao, Sara M Dann, Yingzi Cong
ABSTRACT
Gut bacteria-derived short-chain fatty acids (SCFAs) play crucial roles in the maintenance of intestinal homeostasis. However, how SCFAs regulate epithelial turnover and tissue repair remain incompletely understood. In this study, we investigated how the SCFA propionate regulates cell migration to promote epithelial renewal and repair. Mouse small intestinal epithelial cells (MSIE) and human Caco-2 cells were used to determine the effects of SCFAs on gene expression, proliferation, migration, and cell spreading in vitro. Video microscopy and single cell tracking were used to assess cell migration kinetically. 5-bromo-2'-deoxyuridine (BrdU) and hydroxyurea were used to assess the effects of SCFAs on migration in vivo. Lastly, an acute colitis model using dextran sulfate sodium (DSS) was used to examine the effects of SCFAs in vivo. Using video microscopy and single cell tracking, we found that propionate promoted intestinal epithelial cell migration by enhancing cell spreading and polarization, which led to increases in both cell speed and persistence. This novel function of propionate was dependent on inhibition of class I histone deacetylases (HDAC) and GPR43 and required signal transducer and activator of transcription 3 (STAT3). Furthermore, using 5-bromo-2'-deoxyuridine (BrdU) and hydroxyurea in vivo, we found that propionate enhanced cell migration up the crypt-villus axis under homeostatic conditions, while also protecting against ulcer formation in experimental colitis. Our results demonstrate a mechanism by which propionate stimulates cell migration in an HDAC inhibition, GPR43, and STAT3 dependent manner, and suggest that propionate plays an important role in epithelial migration independent of proliferation.
Sigma-Aldrich
Bz-423, ≥98% (HPLC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Sodium propionate, ≥99.0%
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Supelco
Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium dichloroacetate, 98%
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder