Merck
  • Early B Cell Factor Activity Controls Developmental and Adaptive Thermogenic Gene Programming in Adipocytes.
Early B Cell Factor Activity Controls Developmental and Adaptive Thermogenic Gene Programming in Adipocytes.
Cell reports (2020-03-05)
Anthony R Angueira, Suzanne N Shapira, Jeff Ishibashi, Samay Sampat, Jaimarie Sostre-Colón, Matthew J Emmett, Paul M Titchenell, Mitchell A Lazar, Hee-Woong Lim, Patrick Seale
ABSTRACT
Brown adipose tissue (BAT) activity protects animals against hypothermia and represents a potential therapeutic target to combat obesity. The transcription factor early B cell factor-2 (EBF2) promotes brown adipocyte differentiation, but its roles in maintaining brown adipocyte fate and in stimulating BAT recruitment during cold exposure were unknown. We find that the deletion of Ebf2 in adipocytes of mice ablates BAT character and function, resulting in cold intolerance. Unexpectedly, prolonged exposure to cold restores the thermogenic profile and function of Ebf2 mutant BAT. Enhancer profiling and genetic assays identified EBF1 as a candidate regulator of the cold response in BAT. Adipocyte-specific deletion of both Ebf1 and Ebf2 abolishes BAT recruitment during chronic cold exposure. Mechanistically, EBF1 and EBF2 promote thermogenic gene transcription through increasing the expression and activity of ERRα and PGC1α. Together, these studies demonstrate that EBF proteins specify the developmental fate and control the adaptive cold response of brown adipocytes.
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Roche
Bovine Serum Albumin Fraction V, heat shock, fatty acid free, from bovine serum
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g