Merck
  • Cationic surfactants from lysine: synthesis, micellization and biological evaluation.
Cationic surfactants from lysine: synthesis, micellization and biological evaluation.
European journal of medicinal chemistry (2008-12-17)
Lourdes Pérez, Aurora Pinazo, M Teresa García, Marina Lozano, Angeles Manresa, Marta Angelet, M Pilar Vinardell, Montse Mitjans, Ramon Pons, M Rosa Infante
ABSTRACT
Biocompatible cationic surfactants from the amino acid lysine (hydrochloride salts of N(epsilon)-lauroyl lysine methyl ester, N(epsilon)-myristoyl lysine methyl ester and N(epsilon)-palmitoyl lysine methyl ester) have been prepared in high yields by lysine acylation in epsilon position with three natural saturated fatty acids. The micellization process of these surfactants has been studied using the PGSE-NMR technique. The compounds were tested as antimicrobial agents against Gram-positive and Gram-negative bacteria. The surfactants show moderate antimicrobial activity against the Gram-positive bacteria but Gram-negative bacteria are resistant to these surfactants in the concentration range tested. The haemolytic activity is considerably lower than those reported for other cationic N(alpha)-acyl amino acid analogues. The acute toxicity against Daphnia magna and biodegradability was studied. The toxicity is clearly lower than that reported for conventional cationic surfactants from quaternary ammonium and the three surfactants from lysine can be classified as ready biodegradable surfactants.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Supelco
Hexadecyltrimethylammonium bromide, analytical standard