Merck
  • Agrin induces long-term osteochondral regeneration by supporting repair morphogenesis.
Agrin induces long-term osteochondral regeneration by supporting repair morphogenesis.
Science translational medicine (2020-09-04)
Suzanne E Eldridge, Aida Barawi, Hui Wang, Anke J Roelofs, Magdalena Kaneva, Zeyu Guan, Helen Lydon, Bethan L Thomas, Anne-Sophie Thorup, Beatriz F Fernandez, Sara Caxaria, Danielle Strachan, Ahmed Ali, Kanatheepan Shanmuganathan, Costantino Pitzalis, James R Whiteford, Frances Henson, Andrew W McCaskie, Cosimo De Bari, Francesco Dell'Accio
ABSTRACT
Cartilage loss leads to osteoarthritis, the most common cause of disability for which there is no cure. Cartilage regeneration, therefore, is a priority in medicine. We report that agrin is a potent chondrogenic factor and that a single intraarticular administration of agrin induced long-lasting regeneration of critical-size osteochondral defects in mice, with restoration of tissue architecture and bone-cartilage interface. Agrin attracted joint resident progenitor cells to the site of injury and, through simultaneous activation of CREB and suppression of canonical WNT signaling downstream of β-catenin, induced expression of the chondrogenic stem cell marker GDF5 and differentiation into stable articular chondrocytes, forming stable articular cartilage. In sheep, an agrin-containing collagen gel resulted in long-lasting regeneration of bone and cartilage, which promoted increased ambulatory activity. Our findings support the therapeutic use of agrin for joint surface regeneration.
Sigma-Aldrich
Wnt Agonist II, SKL2001, The Wnt Agonist II, SKL2001 controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Biochemicals applications.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2