Merck
  • Ganaxolone enhances microglial clearance activity and promotes remyelination in focal demyelination in the corpus callosum of ovariectomized rats.
Ganaxolone enhances microglial clearance activity and promotes remyelination in focal demyelination in the corpus callosum of ovariectomized rats.
CNS neuroscience & therapeutics (2019-07-25)
Abdeslam Mouihate, Samah Kalakh
ABSTRACT
Experimental studies have shown that the progesterone metabolite, allopregnanolone, is endowed with promyelinating effects. The mechanisms underlying these promyelinating effects are not well understood. Therefore, we explored the impact of allopregnanolone's synthetic analogue, ganaxolone, on remyelination and microglial activation following focal demyelination in the corpus callosum of ovariectomized rats. Ovariectomized adult Sprague Dawley rats received a stereotaxic injection of 2 µL of 1% lysolecithin solution in the corpus callosum followed by daily injections of either ganaxolone (intraperitoneal injection [i.p.], 2.5 mg/kg) or vehicle. The demyelination lesion was assessed 3 and 7 days postdemyelination insult using Luxol fast blue staining and transmission electron microscopy. The expression levels of myelin proteins (MBP, MAG, MOG, CNPase) were explored using Western blot. The inflammatory response and clearance of damaged myelin were evaluated using immunofluorescent staining (Iba1, dMBP, GFAP) and multiplex enzyme-linked immunosorbent assay (IL-1β, TNF-α, IL-4, IL-10, IL-6). Systemic administration of ganaxolone promoted remyelination of lysolecithin-induced demyelination, upregulated the expression of major myelin proteins, and enhanced microglial clearance of damaged myelin. Astrocytosis, as well as locally produced pro- and antiinflammatory cytokines, was not affected by ganaxolone treatment. Ganaxolone promotes remyelination in response to focal demyelination of the corpus callosum of ovariectomized rats. This effect is, at least in part, mediated by enhancing microglial clearance of myelin debris, which creates a conducive environment for a successful remyelination process.
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Myelin Basic Protein Mouse mAb (SMI-99), liquid, clone SMI-99, Calbiochem®
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Anti- GABAA Antibody γ2 Subunit, clone KC4-8A7, clone KC4-8A7, from mouse