Merck
  • Thbs1 induces lethal cardiac atrophy through PERK-ATF4 regulated autophagy.
Thbs1 induces lethal cardiac atrophy through PERK-ATF4 regulated autophagy.
Nature communications (2021-06-26)
Davy Vanhoutte, Tobias G Schips, Alexander Vo, Kelly M Grimes, Tanya A Baldwin, Matthew J Brody, Federica Accornero, Michelle A Sargent, Jeffery D Molkentin
ABSTRACT
The thrombospondin (Thbs) family of secreted matricellular proteins are stress- and injury-induced mediators of cellular attachment dynamics and extracellular matrix protein production. Here we show that Thbs1, but not Thbs2, Thbs3 or Thbs4, induces lethal cardiac atrophy when overexpressed. Mechanistically, Thbs1 binds and activates the endoplasmic reticulum stress effector PERK, inducing its downstream transcription factor ATF4 and causing lethal autophagy-mediated cardiac atrophy. Antithetically, Thbs1-/- mice develop greater cardiac hypertrophy with pressure overload stimulation and show reduced fasting-induced atrophy. Deletion of Thbs1 effectors/receptors, including ATF6α, CD36 or CD47 does not diminish Thbs1-dependent cardiac atrophy. However, deletion of the gene encoding PERK in Thbs1 transgenic mice blunts the induction of ATF4 and autophagy, and largely corrects the lethal cardiac atrophy. Finally, overexpression of PERK or ATF4 using AAV9 gene-transfer similarly promotes cardiac atrophy and lethality. Hence, we identified Thbs1-mediated PERK-eIF2α-ATF4-induced autophagy as a critical regulator of cardiomyocyte size in the stressed heart.
Sigma-Aldrich
Anti-GRP78/BiP (ET-21) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lectin from Triticum vulgaris (wheat), FITC conjugate, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-SEL1L (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Derlin-3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GRP78/BiP (GL-19) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution