Merck
  • In vitro inhibition of Streptococcus mutans biofilm formation on hydroxyapatite by subinhibitory concentrations of anthraquinones.
In vitro inhibition of Streptococcus mutans biofilm formation on hydroxyapatite by subinhibitory concentrations of anthraquinones.
Antimicrobial agents and chemotherapy (2007-01-16)
Tom Coenye, Kris Honraet, Petra Rigole, Pol Nadal Jimenez, Hans J Nelis
ABSTRACT
We report that certain anthraquinones (AQs) reduce Streptococcus mutans biofilm formation on hydroxyapatite at concentrations below the MIC. Although AQs are known to generate reactive oxygen species, the latter do not underlie the observed effect. Our results suggest that AQs inhibit S. mutans biofilm formation by causing membrane perturbation.
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Physcion, ≥98.0% (TLC)
Sigma-Aldrich
Aloe-emodin, ≥95% (HPLC)
Sigma-Aldrich
Rhein
Sigma-Aldrich
Hypericin from Hypericum perforatum, ~95% (HPLC)
Sigma-Aldrich
Carminic acid, ≥90% (HPLC)
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)