Merck
RADX condenses single-stranded DNA to antagonize RAD51 loading.
Nucleic acids research (2020-07-06)
Hongshan Zhang, Jeffrey M Schaub, Ilya J Finkelstein
ABSTRACT
RADX is a mammalian single-stranded DNA-binding protein that stabilizes telomeres and stalled replication forks. Cellular biology studies have shown that the balance between RADX and Replication Protein A (RPA) is critical for DNA replication integrity. RADX is also a negative regulator of RAD51-mediated homologous recombination at stalled forks. However, the mechanism of RADX acting on DNA and its interactions with RPA and RAD51 are enigmatic. Using single-molecule imaging of the key proteins in vitro, we reveal that RADX condenses ssDNA filaments, even when the ssDNA is coated with RPA at physiological protein ratios. RADX compacts RPA-coated ssDNA filaments via higher-order assemblies that can capture ssDNA in trans. Furthermore, RADX blocks RPA displacement by RAD51 and prevents RAD51 loading on ssDNA. Our results indicate that RADX is an ssDNA condensation protein that inhibits RAD51 filament formation and may antagonize other ssDNA-binding proteins on RPA-coated ssDNA.
Sigma-Aldrich
D-(+)-Maltose monohydrate, ≥98% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Corning® Spin-X® UF concentrators, Spin-X UF 6 for samples up to 6mL; 50K MWCO