Merck
  • Paper-based electrochemical peptide nucleic acid (PNA) biosensor for detection of miRNA-492: a pancreatic ductal adenocarcinoma biomarker.
Paper-based electrochemical peptide nucleic acid (PNA) biosensor for detection of miRNA-492: a pancreatic ductal adenocarcinoma biomarker.
Biosensors & bioelectronics (2020-07-31)
Maria Moccia, Veronica Caratelli, Stefano Cinti, Biagio Pede, Concetta Avitabile, Michele Saviano, Anna Lisa Imbriani, Danila Moscone, Fabiana Arduini
ABSTRACT
Pancreatic ductal adenocarcinoma is the predominant neoplastic disease of the pancreas and it represents the fourth most frequent cause of death in cancer-related disease, with only 8% of survivors after 5-year to the diagnosis. The main issues of this type of cancer rely on fast progress (i.e. 14 months from T1 to a T4 stage), nonspecific symptoms with delay in diagnosis, and the absence of effective screening strategies. To address the lack of early diagnosis, we report a cost-effective paper-based biosensor for the detection of miRNA-492, which is recognised as a biomarker for pancreatic ductal adenocarcinoma. To design a miniaturised, sensitive, and robust paper-based platform, an electrochemical sensor was screen-printed on office paper previously wax-patterned via wax-printing technique. The paper-based sensor was then engineered with a novel and highly specific peptide nucleic acid (PNA) as the recognition element. The formation of PNA/miRNA-492 adduct was evaluated by monitoring the interaction between the positively charged ruthenium (III) hexamine with uncharged PNA and/or negatively charged PNA/miRNA-492 duplex by differential pulse voltammetry. The paper-based biosensor provided a linear range up to 100 nM, with a LOD of 6 nM. Excellent selectivity towards one- and two-base mismatches (1MM, 2MM) or scrambled (SCR) sequences was highlighted and the applicability for biomedical analyses was demonstrated, measuring miRNA-492 in undiluted serum samples.
Sigma-Aldrich
Hexaammineruthenium(III) chloride, 98%
Sigma-Aldrich
2-Methylmorpholine