Merck
Oligomycin-induced proton uncoupling.
Toxicology in vitro : an international journal published in association with BIBRA (2020-06-06)
Abby Hearne, Haotong Chen, Anna Monarchino, Jeffrey S Wiseman
ABSTRACT
Oligomycin is a classical mitochondrial reagent that binds to the proton channel on the Fo component of ATP synthase. As a result, oligomycin blocks mitochondrial ATP synthesis, proton translocation, and O2 uptake. Here we show that oligomycin induces proton uncoupling subsequent to inhibition of ATP synthesis, as evidenced by recovery of O2 uptake to near baseline levels. Uncoupling is uniquely rapid and readily observed in HepG2 cells but is also observed at longer times in the unrelated H1299 cell line. Proton fluxes plateau at oligomycin concentrations in the region 0.25-5 μM. At the plateau, fluxes are lower than expected for the classical mitochondrial permeability transition pore, although in H1229 cells, fluxes increase to levels consistent with pore opening at higher oligomycin concentrations. Uncoupling is observed in cells metabolizing either pyruvate or lactate and reversed by addition of glucose to restore ATP synthesis. Uncoupling is not sensitive to cyclosporin A and is not reversed by the ANT inhibitor bongkrekic acid. However, bongkrekic acid inhibits uncoupling if added before oligomycin, which we interpret in terms of maintenance of mitochondrial ATP levels.
Sigma-Aldrich
Bz-423, ≥98% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Sodium L-lactate, ~98%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
EPPS, ≥99.5% (titration)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Bongkrekic acid solution, from Pseudomonas cocovenenans, ≥95% (HPLC), ~1 mg/mL
Sigma-Aldrich
2,3-Dimethoxy-1,4-naphthoquinone, ≥99%, solid
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture