Merck
  • Real-Time In-Cell NMR Reveals the Intracellular Modulation of GTP-Bound Levels of RAS.
Real-Time In-Cell NMR Reveals the Intracellular Modulation of GTP-Bound Levels of RAS.
Cell reports (2020-08-28)
Qingci Zhao, Ryu Fujimiya, Satoshi Kubo, Christopher B Marshall, Mitsuhiko Ikura, Ichio Shimada, Noritaka Nishida
ABSTRACT
The small guanosine triphosphatase (GTPase) RAS serves as a molecular switch in signal transduction, and its mutation and aberrant activation are implicated in tumorigenesis. Here, we perform real-time, in-cell nuclear magnetic resonance (NMR) analyses of non-farnesylated RAS to measure time courses of the fraction of the active GTP-bound form (fGTP) within cytosol of live mammalian cells. The observed intracellular fGTP is significantly lower than that measured in vitro for wild-type RAS as well as oncogenic mutants, due to both decrease of the guanosine diphosphate (GDP)-GTP exchange rate (kex) and increase of GTP hydrolysis rate (khy). In vitro reconstitution experiments show that highly viscous environments promote a reduction of kex, whereas the increase of khy is stimulated by unidentified cytosolic proteins. This study demonstrates the power of in-cell NMR to directly detect the GTP-bound levels of RAS in mammalian cells, thereby revealing that the khy and kex of RAS are modulated by various intracellular factors.
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Millipore
HIS-Select® Nickel Affinity Gel, (1:1 suspension in a 20% ethanol solution)
Sigma-Aldrich
Guanosine 5′-[γ-thio]triphosphate tetralithium salt, ≥90% (contains < 10% GDP, HPLC), powder
Sigma-Aldrich
Acetate Kinase from Escherichia coli, lyophilized powder, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
Sigma-Aldrich
D-Glucose-1,2,3,4,5,6,6-d7, 97 atom % D