Merck
  • Specific targeting of IL-1β activity to CD8+ T cells allows for safe use as a vaccine adjuvant.
Specific targeting of IL-1β activity to CD8+ T cells allows for safe use as a vaccine adjuvant.
NPJ vaccines (2020-07-28)
Bram Van Den Eeckhout, Lien Van Hoecke, Elianne Burg, Sandra Van Lint, Frank Peelman, Niko Kley, Gilles Uzé, Xavier Saelens, Jan Tavernier, Sarah Gerlo
ABSTRACT
Annual administration and reformulation of influenza vaccines is required for protection against seasonal infections. However, the induction of strong and long-lasting T cells is critical to reach broad and potentially lifelong antiviral immunity. The NLRP3 inflammasome and its product interleukin-1β (IL-1β) are pivotal mediators of cellular immune responses to influenza, yet, overactivation of these systems leads to side effects, which hamper clinical applications. Here, we present a bypass around these toxicities by targeting the activity of IL-1β to CD8+ T cells. Using this approach, we demonstrate safe inclusion of IL-1β as an adjuvant in vaccination strategies, leading to full protection of mice against a high influenza virus challenge dose by raising potent T cell responses. In conclusion, this paper proposes a class of IL-1β-based vaccine adjuvants and also provides further insight in the mechanics of cellular immune responses driven by IL-1β.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Venor GeM Mycoplasma Detection Kit, PCR-based
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, ≥150 IU/mg (dry basis)
Sigma-Aldrich
Sigma Adjuvant System®, oil
Supelco
Timestrip PLUS 8 °C 48 hours