Merck
  • CMTM6 expressed on the adaxonal Schwann cell surface restricts axonal diameters in peripheral nerves.
CMTM6 expressed on the adaxonal Schwann cell surface restricts axonal diameters in peripheral nerves.
Nature communications (2020-09-11)
Maria A Eichel, Vasiliki-Ilya Gargareta, Elisa D'Este, Robert Fledrich, Theresa Kungl, Tobias J Buscham, Katja A Lüders, Cristina Miracle, Ramona B Jung, Ute Distler, Kathrin Kusch, Wiebke Möbius, Swen Hülsmann, Stefan Tenzer, Klaus-Armin Nave, Hauke B Werner
ABSTRACT
The velocity of nerve conduction is moderately enhanced by larger axonal diameters and potently sped up by myelination of axons. Myelination thus allows rapid impulse propagation with reduced axonal diameters; however, no myelin-dependent mechanism has been reported that restricts radial growth of axons. By label-free proteomics, STED-microscopy and cryo-immuno electron-microscopy we here identify CMTM6 (chemokine-like factor-like MARVEL-transmembrane domain-containing family member-6) as a myelin protein specifically localized to the Schwann cell membrane exposed to the axon. We find that disruption of Cmtm6-expression in Schwann cells causes a substantial increase of axonal diameters but does not impair myelin biogenesis, radial sorting or integrity of axons. Increased axonal diameters correlate with accelerated sensory nerve conduction and sensory responses and perturbed motor performance. These data show that Schwann cells utilize CMTM6 to restrict the radial growth of axons, which optimizes nerve function.
Sigma-Aldrich
Anti-TUBA4A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid