Merck
  • Biochemistry and biomedicine of quantum dots: from biodetection to bioimaging, drug discovery, diagnostics, and therapy.
Biochemistry and biomedicine of quantum dots: from biodetection to bioimaging, drug discovery, diagnostics, and therapy.
Acta biomaterialia (2018-05-08)
Jun Yao, Pingfan Li, Lin Li, Mei Yang
ABSTRACT
According to recent research, nanotechnology based on quantum dots (QDs) has been widely applied in the field of bioimaging, drug delivery, and drug analysis. Therefore, it has become one of the major forces driving basic and applied research. The application of nanotechnology in bioimaging has been of concern. Through in vitro labeling, it was found that luminescent QDs possess many properties such as narrow emission, broad UV excitation, bright fluorescence, and high photostability. The QDs also show great potential in whole-body imaging. The QDs can be combined with biomolecules, and hence, they can be used for targeted drug delivery and diagnosis. The characteristics of QDs make them useful for application in pharmacy and pharmacology. This review focuses on various applications of QDs, especially in imaging, drug delivery, pharmaceutical analysis, photothermal therapy, biochips, and targeted surgery. Finally, conclusions are made by providing some critical challenges and a perspective of how this field can be expected to develop in the future. Quantum dots (QDs) is an emerging field of interdisciplinary subject that involves physics, chemistry, materialogy, biology, medicine, and so on. In addition, nanotechnology based on QDs has been applied in depth in biochemistry and biomedicine. Some forward-looking fields emphatically reflected in some extremely vital areas that possess inspiring potential applicable prospects, such as immunoassay, DNA analysis, biological monitoring, drug discovery, in vitro labelling, in vivo imaging, and tumor target are closely connected to human life and health and has been the top and forefront in science and technology to date. Furthermore, this review has not only involved the traditional biochemical detection but also particularly emphasized its potential applications in life science and biomedicine.
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 540 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 470 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 450 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 520 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 470 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 580 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 600 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 500 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 520 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 540 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 650 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 600 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λex 540 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 620 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 560 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λex 650 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 580 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 600 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 620 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 620 nm, 5 mg/mL in toluene
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 520 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 500 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 560 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 470 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 500 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 560 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 650 nm, 1 mg/mL in water
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, oleic acid functionalized, fluorescence λem 580 nm, 5 mg/mL in toluene