Merck
  • High-efficiency inverted polymer solar cells with transparent and work-function tunable MoO(3)-Al composite film as cathode buffer layer.
High-efficiency inverted polymer solar cells with transparent and work-function tunable MoO(3)-Al composite film as cathode buffer layer.
Advanced materials (Deerfield Beach, Fla.) (2012-04-19)
Jian Liu, Shuyan Shao, Gang Fang, Bin Meng, Zhiyuan Xie, Lixiang Wang
ABSTRACT
High-efficiency inverted polymer solar cells based on PCDTBT:PC(70)BM blend with the MoO(3)-Al composite film as the cathode buffer layer and the MoO(3)/Al as the anode have been demonstrated. A V(OC) of 0.88 V, a J(SC) of 10.88 mA cm(-2), a FF of 70.7% and a PCE of 6.77% are achieved. The MoO(3)-Al composite films are highly transparent with adjustable work functions which can be fine tuned based on the Al content in the composite, thus allowing us to optimize the interfacial property at cathode buffer layer/BHJ interfaces to reduce recombination loss and to improve the photovoltaic performance. This new approach has simplified the device fabrication and will render economizing in large scale applications.
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis