Merck
  • Near-Infrared-Activated Fluorescence Resonance Energy Transfer-Based Nanocomposite to Sense MMP2-Overexpressing Oral Cancer Cells.
Near-Infrared-Activated Fluorescence Resonance Energy Transfer-Based Nanocomposite to Sense MMP2-Overexpressing Oral Cancer Cells.
ACS omega (2018-07-20)
Yung-Chieh Chan, Ming-Hsien Chan, Chieh-Wei Chen, Ru-Shi Liu, Michael Hsiao, Din Ping Tsai
ABSTRACT
The matrix metalloproteinases (MMPs) are well-known mediators that are activated in tumor progression. MMP2 is a kind of gelatinase in extracellular matrix remodeling and cancer metastasis processes. MMP2 secretion increased in many types of cancer diseases, and its abnormal expression is associated with a poor prognosis. We fabricated a nanocomposite that sensed MMP2 expression by a red and blue light change. This nanocomposite consisted of an upconversion nanoparticle (UCNP), MMP2-sensitive peptide, and CuInS2/ZnS quantum dot (CIS/ZnS QD). An UCNP is composed of NaYF4:Tm/Yb@NaYF4:Nd/Yb, which has multiple emissions at UV/blue-visible wavelengths under 808 nm laser excitation. The conjugated CIS/ZnS QD showed the red-visible fluorescence though the FRET process. The two fluorophores were connected by a MMP2-sensitive peptide to form a novel MMP2 biosensor, named UCNP@p-QD. UCNP@p-QD was highly biocompatible according to cell viability assay. The FRET-based biosensor was employed in the MMP2 determination in vitro and in vivo. Furthermore, it was administrated into the tumor-bearing mouse to check MMP2 expression. UCNP@p-QD could be a promising tool for biological study and biomedical application. In this study, we demonstrated that the CIS/ZnS QD improved the upconversion intensity through a near-infrared-induced FRET process. This nanocomposite has the advantage of light penetration, excellent biocompatibility, and high sensitivity to sense MMP2. The near-infrared-induced composites are a potential inspiration for use in biomedical applications.
Sigma-Aldrich
m-dPEG®4-thiol, >95% (HPLC)
Sigma-Aldrich
Thiol-dPEG®12-acid, >95% (HPLC)