Merck
  • Proximate Analysis and Nutritional Evaluation of Twenty Canadian Lentils by Principal Component and Cluster Analyses.
Proximate Analysis and Nutritional Evaluation of Twenty Canadian Lentils by Principal Component and Cluster Analyses.
Foods (Basel, Switzerland) (2020-02-15)
D Dan Ramdath, Zhan-Hui Lu, Padma L Maharaj, Jordan Winberg, Yolanda Brummer, Aileen Hawke
ABSTRACT
Proximate composition and starch nutritional properties of twenty cooked lentils were assessed to identify unique varieties that could be used in value added foods. Significant variations exist among the lentil varieties (p < 0.05) with respect to their energy, fat, protein, carbohydrate, and dietary fiber content, and these are related to lentil type and seed size. Dazil and Greenstar were unique for their high resistant starch content (RS) and lower area under the starch hydrolysis curve (SHAUC) while Proclaim was opposite. SHAUC was positively correlated (p < 0.001) with rapidly digestible starch (RDS) content (r = 0.626) but negatively correlated with RS content (r = -0.635). Principal component analysis showed that the first three principal components accounted for 62.8% of the total variance and the contribution of SHAUC was 33.2%. These results confirm that in vitro SHAUC and a combination of RDS and RS may be predictive of the digestibility profile of cooked lentils.
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Starch from corn