Merck
  • Frozen dog spermatozoa are negatively affected during storage at -80, -21 and -8 ºC.
Frozen dog spermatozoa are negatively affected during storage at -80, -21 and -8 ºC.
Animal reproduction science (2019-10-23)
Antonio González, Ander Arando, Alberto Acosta, Carlos J Alcalá, Francisco A Arrebola, Carlos C Pérez-Marín
ABSTRACT
Cryopreservation in liquid nitrogen (LN2) allows for semen to be stored for long periods of time while there is sustaining of sperm viability. In this study, there was assessment of effects induced by different storage temperatures on cryopreserved dog spermatozoa. After cryopreservation at -196 °C, sperm samples were transferred to storage conditions of -80, 21 or -8 °C. Sperm motility, morphology, viability, acrosome integrity, mitochondrial membrane potential and DNA fragmentation were determined in samples stored at -196 °C (evaluation time =0 h), and then after 12 h and 1, 4, 7 and 15 d of storage at 80, -21 and -8 °C. In samples stored at -80 °C, sperm morphology, viability, acrosome integrity, mitochondrial membrane potential and DNA fragmentation did not differ at successive evaluation times. Progressive motility was less (P < 0.05) after 12 h and total motility after 4 d of storage at -80 ºC as compared with that of the 0 h sample. With storage at the other temperatures (-21 and -8 ºC), there was a reduction of mean values for sperm total and progressive motility, viability and mitochondrial membrane potential after 12 h of storage at these temperatures. Results, therefore, indicate the use of ultra-freezers at -80 ºC to store frozen dog semen allows for maintenance of sperm characteristics for at least 15 d but motility is sustained for only 1 d. Neither of the -21 or -8 ºC storage temperatures were effective for storing of frozen dog sperm and retaining viability.
Product Description
Pyrex® Lifetime Red graduated tube, O.D. × L 25 mm × 200 mm, volume 50.0 mL, accuracy: 0.4 mL