Merck
  • One-Pot Generation of Benzynes from Phenols: Formation of Primary Anilines by the Deoxyamination of Phenols.
One-Pot Generation of Benzynes from Phenols: Formation of Primary Anilines by the Deoxyamination of Phenols.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-01-03)
Takashi Ikawa, Shigeaki Masuda, Shuji Akai
ABSTRACT
Benzynes were selectively generated in situ from phenols and trapped regioselectively with potassium hexamethyldisilazide to form primary anilines following acidic workup. The direct conversion of a phenolic hydroxyl group into a free amino group is a useful method for the preparation of primary aryl amines that are hard to synthesize by using coupling reactions involving phenol derivatives with ammonia. Whereas reactions of ortho- and meta-substituted phenols produced meta-substituted anilines exclusively, those of para-substituted phenols provided ortho-silylanilines.
Sigma-Aldrich
3,4-Dimethoxyaniline, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2-(Methylthio)aniline, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
3-(Methylthio)aniline, 97%
Pricing and availability is not currently available.