Merck
  • PRMT1 Is Recruited via DNA-PK to Chromatin Where It Sustains the Senescence-Associated Secretory Phenotype in Response to Cisplatin.
PRMT1 Is Recruited via DNA-PK to Chromatin Where It Sustains the Senescence-Associated Secretory Phenotype in Response to Cisplatin.
Cell reports (2020-01-30)
Daniele Musiani, Roberto Giambruno, Enrico Massignani, Marica Rosaria Ippolito, Marianna Maniaci, Sriganesh Jammula, Daria Manganaro, Alessandro Cuomo, Luciano Nicosia, Diego Pasini, Tiziana Bonaldi
ABSTRACT
Protein arginine methyltransferase 1 (PRMT1) is overexpressed in various human cancers and linked to poor response to chemotherapy. Various PRMT1 inhibitors are currently under development; yet, we do not fully understand the mechanisms underpinning PRMT1 involvement in tumorigenesis and chemoresistance. Using mass spectrometry-based proteomics, we identified PRMT1 as regulator of arginine methylation in ovarian cancer cells treated with cisplatin. We showed that DNA-dependent protein kinase (DNA-PK) binds to and phosphorylates PRMT1 in response to cisplatin, inducing its chromatin recruitment and redirecting its enzymatic activity toward Arg3 of histone H4 (H4R3). On chromatin, the DNA-PK/PRMT1 axis induces senescence-associated secretory phenotype through H4R3me2a deposition at pro-inflammatory gene promoters. Finally, PRMT1 inhibition reduces the clonogenic growth of cancer cells exposed to low doses of cisplatin, sensitizing them to apoptosis. While unravelling the role of PRMT1 in response to genotoxic agents, our findings indicate the possibility of targeting PRMT1 to overcome chemoresistance in cancer.
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
L-Methionine-(methyl-13C,d3), ≥99 atom % 13C, ≥99 atom % D, ≥99% (CP)
Sigma-Aldrich
Bay 11-7085, ≥98% (HPLC), solid
Sigma-Aldrich
L-Lysine-4,4,5,5-d4 hydrochloride, 98 atom % D, 98% (CP)
Sigma-Aldrich
L-Arginine-13C6,15N4 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
L-Arginine-13C6 hydrochloride, 99 atom % 13C, 95% (CP)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Anti-dimethyl-Histone H4 (Arg3) Asymmetric Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®