Merck
  • Probiotic Bacillus subtilis Protects against α-Synuclein Aggregation in C. elegans.
Probiotic Bacillus subtilis Protects against α-Synuclein Aggregation in C. elegans.
Cell reports (2020-01-16)
María Eugenia Goya, Feng Xue, Cristina Sampedro-Torres-Quevedo, Sofia Arnaouteli, Lourdes Riquelme-Dominguez, Andrés Romanowski, Jack Brydon, Kathryn L Ball, Nicola R Stanley-Wall, Maria Doitsidou
ABSTRACT
Recent discoveries have implicated the gut microbiome in the progression and severity of Parkinson's disease; however, how gut bacteria affect such neurodegenerative disorders remains unclear. Here, we report that the Bacillus subtilis probiotic strain PXN21 inhibits α-synuclein aggregation and clears preformed aggregates in an established Caenorhabditis elegans model of synucleinopathy. This protection is seen in young and aging animals and is partly mediated by DAF-16. Multiple B. subtilis strains trigger the protective effect via both spores and vegetative cells, partly due to a biofilm formation in the gut of the worms and the release of bacterial metabolites. We identify several host metabolic pathways differentially regulated in response to probiotic exposure, including sphingolipid metabolism. We further demonstrate functional roles of the sphingolipid metabolism genes lagr-1, asm-3, and sptl-3 in the anti-aggregation effect. Our findings provide a basis for exploring the disease-modifying potential of B. subtilis as a dietary supplement.
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
MAHMA NONOate
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Iron(II) sulfate hydrate, 99.999% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Supelco
Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 100 g
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, free-flowing, Redi-Dri, ≥98%
Sigma-Aldrich
Calcium nitrate tetrahydrate, ≥99.0%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%