- Hydrogen adsorption in the nanoporous metal-benzenedicarboxylate M(OH)(O2C-C6H4-CO2) (M = Al3+, Cr3+), MIL-53.
Hydrogen adsorption in the nanoporous metal-benzenedicarboxylate M(OH)(O2C-C6H4-CO2) (M = Al3+, Cr3+), MIL-53.
Chemical communications (Cambridge, England) (2004-01-06)
Gérard Férey, Michel Latroche, Christian Serre, Franck Millange, Thierry Loiseau, Annick Percheron-Guégan
PMID14703815
ABSTRACT
Hydrogen adsorption has been studied in the nanoporous metal-benzenedicarboxylate M(OH)(O2C-C6H4-CO2) (M = Al3+, Cr3+); these solids show a hydrogen storage capacity of 3.8 and 3.1 wt.% respectively when loaded at 77 K under 1.6 MPa.