Merck
  • Epididymal protein Rnase10 is required for post-testicular sperm maturation and male fertility.
Epididymal protein Rnase10 is required for post-testicular sperm maturation and male fertility.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2012-07-04)
Anton Krutskikh, Ariel Poliandri, Victoria Cabrera-Sharp, Jean Louis Dacheux, Matti Poutanen, Ilpo Huhtaniemi
ABSTRACT
Eutherian spermatozoa are dependent on the environment of the proximal epididymis to complete their maturation; however, no specific epididymal factors that mediate this process have so far been identified. Here, we show that targeted disruption of the novel gene Rnase10 encoding a secreted proximal epididymal protein in the mouse results in a binding defect in spermatozoa and their inability to pass through the uterotubal junction in the female. The failure to gain the site of fertilization in the knockout spermatozoa is associated with a gradual loss of ADAM3 and ADAM6 proteins during epididymal transit. In the distal epididymis, these spermatozoa appear to lack calcium-dependent associations with the immobilizing glutinous extracellular material and are released as single, vigorously motile cells that display no tendency for head-to-head agglutination and lack affinity to the oviductal epithelium. In sperm-egg binding assay, they are unable to establish a tenacious association with the zona pellucida, yet they are capable of fertilization. Furthermore, these sperm show accelerated capacitation resulting in an overall in vitro fertilizing ability superior to that of wild-type sperm. We conclude that the physiological role of sperm adhesiveness is in the mechanism of restricted sperm entry into the oviduct rather than in sperm-egg interaction.
Sigma-Aldrich
Anti-ADAM2 Antibody, clone 9D2.2, clone 9D2.2, Chemicon®, from mouse