Merck
  • Blockade of IDO-kynurenine-AhR metabolic circuitry abrogates IFN-γ-induced immunologic dormancy of tumor-repopulating cells.
Blockade of IDO-kynurenine-AhR metabolic circuitry abrogates IFN-γ-induced immunologic dormancy of tumor-repopulating cells.
Nature communications (2017-05-11)
Yuying Liu, Xiaoyu Liang, Xiaonan Yin, Jiadi Lv, Ke Tang, Jingwei Ma, Tiantian Ji, Huafeng Zhang, Wenqian Dong, Xun Jin, Degao Chen, Yanchun Li, Songyan Zhang, Heidi Q Xie, Bin Zhao, Tong Zhao, Jinzhi Lu, Zhuo-Wei Hu, Xuetao Cao, F Xiao-Feng Qin, Bo Huang
ABSTRACT
Interactions with the immune system may lead tumorigenic cells into dormancy. However, the underlying molecular mechanism is poorly understood. Using a 3D fibrin gel model, we show that IFN-γ induces tumour-repopulating cells (TRCs) to enter dormancy through an indolamine 2,3-dioxygenase 1 (IDO1)-kynurenine (Kyn)-aryl hydrocarbon receptor (AhR)-p27 dependent pathway. Mechanistically, IFN-γ signalling triggers differentiated tumour cell apoptosis via STAT1; however, when IDO1 and AhR are highly expressed as in TRCs, IFN-γ results in IDO1/AhR-dependent p27 induction that prevents STAT1 signalling, thus suppressing the process of cell death and activating the dormancy program. Blocking the IDO/AhR metabolic circuitry not only abrogates IFN-γ-induced dormancy but also results in enhanced repression of tumour growth by IFN-γ-induced apoptosis of TRCs both in vitro and in vivo. These data present a previously unrecognized mechanism of inducing TRC dormancy by IFN-γ, suggesting a potential effective cancer immunotherapeutic modality through the combination of IFN-γ and IDO/AhR inhibitors.
Sigma-Aldrich
Anti-IDO-1 Antibody, clone 4B7 | MABF850, clone 4B7, from mouse
Sigma-Aldrich
Rabbit anti-TopBP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken