Merck
  • Naringenin Promotes Thermogenic Gene Expression in Human White Adipose Tissue.
Naringenin Promotes Thermogenic Gene Expression in Human White Adipose Tissue.
Obesity (Silver Spring, Md.) (2018-12-07)
Candida J Rebello, Frank L Greenway, Frank H Lau, Yuan Lin, Jacqueline M Stephens, William D Johnson, Ann A Coulter
ABSTRACT
Naringenin, a citrus flavonoid, prevents diet-induced weight gain and improves glucose and lipid metabolism in rodents. There is evidence that naringenin activates brown fat and increases energy expenditure in mice, but little is known about its effects in humans. The goal of this study was to examine the effects of naringenin on energy expenditure in adipose tissue. Human white adipocyte cultures (hADSC) and abdominal subcutaneous adipose tissue (pWAT) were treated with naringenin for 7 to 14 days. Expression (quantitative real-time polymerase chain reaction, immunoblotting) of candidate genes involved in thermogenesis and glucose metabolism was measured. Oxygen consumption rate was measured in hADSC using a Seahorse flux analyzer. In hADSC, naringenin increased expression of the genes associated with thermogenesis and fat oxidation, including uncoupling protein 1 and adipose triglyceride lipase, and key factors associated with insulin sensitivity, including glucose transporter type 4, adiponectin, and carbohydrate-responsive element-binding protein (P < 0.01). Similar responses were observed in pWAT. Basal, ATP-linked, maximal and reserve oxygen consumption rate increased in the naringenin-treated hADSC (P < 0.01). Naringenin increases energy expenditure in hADSC and stimulates expression of key enzymes involved in thermogenesis and insulin sensitivity in hADSC and pWAT. Naringenin may promote conversion of human white adipose tissue to a brown/beige phenotype.
Sigma-Aldrich
Goat Anti-Mouse IgG & IgM Antibody, HRP conjugate, 0.8 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid