Merck
  • Sonic Hedgehog repression underlies gigaxonin mutation-induced motor deficits in giant axonal neuropathy.
Sonic Hedgehog repression underlies gigaxonin mutation-induced motor deficits in giant axonal neuropathy.
The Journal of clinical investigation (2019-09-11)
Yoan Arribat, Karolina S Mysiak, Léa Lescouzères, Alexia Boizot, Maxime Ruiz, Mireille Rossel, Pascale Bomont
ABSTRACT
Growing evidence shows that alterations occurring at early developmental stages contribute to symptoms manifested in adulthood in the setting of neurodegenerative diseases. Here, we studied the molecular mechanisms causing giant axonal neuropathy (GAN), a severe neurodegenerative disease due to loss-of-function of the gigaxonin-E3 ligase. We showed that gigaxonin governs Sonic Hedgehog (Shh) induction, the developmental pathway patterning the dorso-ventral axis of the neural tube and muscles, by controlling the degradation of the Shh-bound Patched receptor. Similar to Shh inhibition, repression of gigaxonin in zebrafish impaired motor neuron specification and somitogenesis and abolished neuromuscular junction formation and locomotion. Shh signaling was impaired in gigaxonin-null zebrafish and was corrected by both pharmacological activation of the Shh pathway and human gigaxonin, pointing to an evolutionary-conserved mechanism regulating Shh signaling. Gigaxonin-dependent inhibition of Shh activation was also demonstrated in primary fibroblasts from patients with GAN and in a Shh activity reporter line depleted in gigaxonin. Our findings establish gigaxonin as a key E3 ligase that positively controls the initiation of Shh transduction, and reveal the causal role of Shh dysfunction in motor deficits, thus highlighting the developmental origin of GAN.
Product Description
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Cyclopamine hydrate, ≥98% (HPLC)
Sigma-Aldrich
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, purum, ≥98.0% (TLC)
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®