Merck
  • Acute Cocaine Enhances Dopamine D2R Recognition and Signaling and Counteracts D2R Internalization in Sigma1R-D2R Heteroreceptor Complexes.
Acute Cocaine Enhances Dopamine D2R Recognition and Signaling and Counteracts D2R Internalization in Sigma1R-D2R Heteroreceptor Complexes.
Molecular neurobiology (2019-04-12)
Dasiel O Borroto-Escuela, Manuel Narváez, Wilber Romero-Fernández, Luca Pinton, Karolina Wydra, Malgorzata Filip, Sarah Beggiato, Sergio Tanganelli, Luca Ferraro, Kjell Fuxe
ABSTRACT
The current study was performed to establish the actions of nanomolar concentrations of cocaine, not blocking the dopamine transporter, on dopamine D2 receptor (D2R)-sigma 1 receptor (δ1R) heteroreceptor complexes and the D2R protomer recognition, signaling and internalization in cellular models. We report the existence of D2R-δ1R heteroreceptor complexes in subcortical limbic areas as well as the dorsal striatum, with different distribution patterns using the in situ proximity ligation assay. Also, through BRET, these heteromers were demonstrated in HEK293 cells. Furthermore, saturation binding assay demonstrated that in membrane preparations of HEK293 cells coexpressing D2R and δ1R, cocaine (1 nM) significantly increased the D2R Bmax values over cells singly expressing D2R. CREB reporter luc-gene assay indicated that coexpressed δ1R significantly reduced the potency of the D2R-like agonist quinpirole to inhibit via D2R activation the forskolin induced increase of the CREB signal. In contrast, the addition of 100 nM cocaine was found to markedly increase the quinpirole potency to inhibit the forskolin-induced increase of the CREB signal in the D2R-δ1R cells. These events were associated with a marked reduction of cocaine-induced internalization of D2R protomers in D2R-δ1R heteromer-containing cells vs D2R singly expressing cells as studied by means of confocal analysis of D2R-δ1R trafficking and internalization. Overall, the formation of D2R-δ1R heteromers enhanced the ability of cocaine to increase the D2R protomer function associated with a marked reduction of its internalization. The existence of D2R-δ1R heteromers opens up a new understanding of the acute actions of cocaine.
Sigma-Aldrich
Cocaine free base
Sigma-Aldrich
Anti-Dopamine D2 receptor (DRD2) Antibody, clone 2B9, clone 3D9, from mouse