Merck
  • Membrane-associated periodic skeleton is a signaling platform for RTK transactivation in neurons.
Membrane-associated periodic skeleton is a signaling platform for RTK transactivation in neurons.
Science (New York, N.Y.) (2019-08-31)
Ruobo Zhou, Boran Han, Chenglong Xia, Xiaowei Zhuang
ABSTRACT
Actin, spectrin, and related molecules form a membrane-associated periodic skeleton (MPS) in neurons. The function of the MPS, however, remains poorly understood. Using super-resolution imaging, we observed that G protein-coupled receptors (GPCRs), cell adhesion molecules (CAMs), receptor tyrosine kinases (RTKs), and related signaling molecules were recruited to the MPS in response to extracellular stimuli, resulting in colocalization of these molecules and RTK transactivation by GPCRs and CAMs, giving rise to extracellular signal-regulated kinase (ERK) signaling. Disruption of the MPS prevented such molecular colocalizations and downstream ERK signaling. ERK signaling in turn caused calpain-dependent MPS degradation, providing a negative feedback that modulates signaling strength. These results reveal an important functional role of the MPS and establish it as a dynamically regulated platform for GPCR- and CAM-mediated RTK signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human CLTC
Sigma-Aldrich
GNF 5837, ≥98% (HPLC)
Sigma-Aldrich
Phorbol 12,13-dibutyrate, ≥98% (TLC), powder
Sigma-Aldrich
Trypsin solution from porcine pancreas, sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in 0.9% sodium chloride
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
PD 173074, ≥96% (HPLC), powder
Sigma-Aldrich
PP2, ≥98% (HPLC)