Merck
  • RAD51AP1 Is an Essential Mediator of Alternative Lengthening of Telomeres.
RAD51AP1 Is an Essential Mediator of Alternative Lengthening of Telomeres.
Molecular cell (2019-08-12)
Jonathan Barroso-González, Laura García-Expósito, Song My Hoang, Michelle L Lynskey, Justin L Roncaioli, Arundhati Ghosh, Callen T Wallace, Marco de Vitis, Mauro Modesti, Kara A Bernstein, Saumendra N Sarkar, Simon C Watkins, Roderick J O'Sullivan
ABSTRACT
Alternative lengthening of telomeres (ALT) is a homology-directed repair (HDR) mechanism of telomere elongation that controls proliferation in aggressive cancers. We show that the disruption of RAD51-associated protein 1 (RAD51AP1) in ALT+ cancer cells leads to generational telomere shortening. This is due to RAD51AP1's involvement in RAD51-dependent homologous recombination (HR) and RAD52-POLD3-dependent break induced DNA synthesis. RAD51AP1 KO ALT+ cells exhibit telomere dysfunction and cytosolic telomeric DNA fragments that are sensed by cGAS. Intriguingly, they activate ULK1-ATG7-dependent autophagy as a survival mechanism to mitigate DNA damage and apoptosis. Importantly, RAD51AP1 protein levels are elevated in ALT+ cells due to MMS21 associated SUMOylation. Mutation of a single SUMO-targeted lysine residue perturbs telomere dynamics. These findings indicate that RAD51AP1 is an essential mediator of the ALT mechanism and is co-opted by post-translational mechanisms to maintain telomere length and ensure proliferation of ALT+ cancer cells.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-ATM antibody,Mouse monoclonal, clone MAT3-4G10/8, purified from hybridoma cell culture
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Millipore
Benzonase® Nuclease, Purity > 90%