Merck
  • YAP Partially Reprograms Chromatin Accessibility to Directly Induce Adult Cardiogenesis In Vivo.
YAP Partially Reprograms Chromatin Accessibility to Directly Induce Adult Cardiogenesis In Vivo.
Developmental cell (2019-02-19)
Tanner O Monroe, Matthew C Hill, Yuka Morikawa, John P Leach, Todd Heallen, Shuyi Cao, Peter H L Krijger, Wouter de Laat, Xander H T Wehrens, George G Rodney, James F Martin
ABSTRACT
Specialized adult somatic cells, such as cardiomyocytes (CMs), are highly differentiated with poor renewal capacity, an integral reason underlying organ failure in disease and aging. Among the least renewable cells in the human body, CMs renew approximately 1% annually. Consistent with poor CM turnover, heart failure is the leading cause of death. Here, we show that an active version of the Hippo pathway effector YAP, termed YAP5SA, partially reprograms adult mouse CMs to a more fetal and proliferative state. One week after induction, 19% of CMs that enter S-phase do so twice, CM number increases by 40%, and YAP5SA lineage CMs couple to pre-existing CMs. Genomic studies showed that YAP5SA increases chromatin accessibility and expression of fetal genes, partially reprogramming long-lived somatic cells in vivo to a primitive, fetal-like, and proliferative state.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-PCM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic