Merck
  • Integrin alpha4beta1-VCAM-1-mediated adhesion between endothelial and mural cells is required for blood vessel maturation.
Integrin alpha4beta1-VCAM-1-mediated adhesion between endothelial and mural cells is required for blood vessel maturation.
The Journal of clinical investigation (2005-05-20)
Barbara Garmy-Susini, Hui Jin, Yuhong Zhu, Rou-Jia Sung, Rosa Hwang, Judy Varner
ABSTRACT
Neovascularization depends on vascular cell proliferation and on the stabilization of vessels by association of vascular smooth muscle-like pericytes with ECs. Here we show that integrin alpha4beta1 (VLA-4) and VCAM-1 promote close intercellular adhesion between ECs and pericytes and that this interaction is required for blood vessel formation. Integrin alpha4beta1 is expressed by proliferating but not quiescent ECs, while its ligand VCAM-1 is expressed by proliferating but not quiescent mural cells. Antagonists of this integrin-ligand pair block the adhesion of mural cells to proliferating endothelia in vitro and in vivo, thereby inducing apoptosis of ECs and pericytes and inhibiting neovascularization. These studies indicate that integrin alpha4beta1 and VCAM-1 facilitate a critical cell-cell adhesion event required for survival of endothelial and mural cells during vascularization.
Sigma-Aldrich
Anti-VCAM-1 Antibody, clone P8B1, clone P8B1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P4G9, ascites fluid, clone P4G9, Chemicon®
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone HP2/1, clone HP2/1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse