Merck
  • Regulation of the Hippo Pathway by Phosphatidic Acid-Mediated Lipid-Protein Interaction.
Regulation of the Hippo Pathway by Phosphatidic Acid-Mediated Lipid-Protein Interaction.
Molecular cell (2018-10-09)
Han Han, Ruxi Qi, Jeff Jiajing Zhou, Albert Paul Ta, Bing Yang, Hiroki J Nakaoka, Gayoung Seo, Kun-Liang Guan, Ray Luo, Wenqi Wang
ABSTRACT
The Hippo pathway plays a crucial role in organ size control and tumor suppression, but its precise regulation is not fully understood. In this study, we discovered that phosphatidic acid (PA)-related lipid signaling is a key regulator of the Hippo pathway. Supplementing PA in various Hippo-activating conditions activates YAP. This PA-related lipid signaling is involved in Rho-mediated YAP activation. Mechanistically, PA directly interacts with Hippo components LATS and NF2 to disrupt LATS-MOB1 complex formation and NF2-mediated LATS membrane translocation and activation, respectively. Inhibition of phospholipase D (PLD)-dependent PA production suppresses YAP oncogenic activities. PLD1 is highly expressed in breast cancer and positively correlates with YAP activation, suggesting their pathological relevance in breast cancer development. Taken together, our study not only reveals a role of PLD-PA lipid signaling in regulating the Hippo pathway but also indicates that the PLD-PA-YAP axis is a potential therapeutic target for cancer treatment.
Sigma-Aldrich
CI 976, >98% (HPLC), solid
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
FIPI hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture