Merck
  • Investigation of mycobacteria fatty acid profile using different ionization energies in GC-MS.
Investigation of mycobacteria fatty acid profile using different ionization energies in GC-MS.
Analytical and bioanalytical chemistry (2018-10-30)
Marco Beccaria, Flavio A Franchina, Mavra Nasir, Theodore Mellors, Jane E Hill, Giorgia Purcaro
ABSTRACT
Gas chromatography (GC) coupled with electron ionization (EI) mass spectrometry (MS) is a well-established technique for the analysis of volatile and semi-volatile compounds. The main advantage is the highly repeatable fragmentation of the compounds into the ion source, generating intense and diagnostic fragmentation when the ionization is performed at 70 eV; this is considered the standard ionization condition and has been used for creating many established databases, which are of great support in the analyte identification process. However, such an intense fragmentation often causes the loss of the molecular ion or more diagnostic ions, which can be detrimental for the identification of homologous series or isomers, as for instance fatty acids. To obtain this information chemical or soft ionization can be used, but dedicated ion sources and conditions are required. In this work, we explored different ionization voltages in GC-EI-MS to preserve the intensity of the molecular ion using a conventional quadrupole MS. Twenty, 30, 50, and 70 eV were tested using a mixture of fatty acid methyl esters standards. Intensity and repeatability of the most informative ions were compared. Twenty and 70 eV were then used to analyze the fatty acid composition of six different strains of mycobacteria. Two approaches were used for elaborating the data: (1) a single average spectrum of the entire chromatogram was derived, which can be considered (in terms of concept) as a direct EI-MS analysis; (2) the actual chromatographic separation of the compounds was considered after automatic alignment. The results obtained are discussed herein. Graphical abstract ᅟ.
Product Description
Supelco
Trimethylsulfonium hydroxide solution, ~0.25 M in methanol, LiChropur