Merck
  • Inhibition of peptidylarginine deiminase alleviates LPS-induced pulmonary dysfunction and improves survival in a mouse model of lethal endotoxemia.
Inhibition of peptidylarginine deiminase alleviates LPS-induced pulmonary dysfunction and improves survival in a mouse model of lethal endotoxemia.
European journal of pharmacology (2018-07-08)
Yingjian Liang, Baihong Pan, Hasan B Alam, Qiufang Deng, Yibing Wang, Eric Chen, Baoling Liu, Yuzi Tian, Aaron M Williams, Xiuzhen Duan, Yanming Wang, Jifeng Zhang, Yongqing Li
ABSTRACT
Immune cell death caused by neutrophil extracellular traps (NETs), referred to as NETosis, can contribute to the pathogenesis of endotoxemia and organ damage. Although the mechanisms by which infection induces NETosis and how that leads to organ dysfunction remain largely unknown, NET formation is often found following citrullination of histone H3 (CitH3) by peptidylarginine deiminase (PAD). We hypothesized that lipopolysaccharide (LPS)-induced activation of PAD and subsequent CitH3-mediated NET formation increases endothelial permeability and pulmonary dysfunction and, therefore, that inhibition of PAD can mitigate damage and improve survival in lethal endotoxemia. Here, we showed that treatment with YW3-56, a PAD2/PAD4 inhibitor, significantly diminished PAD activation, blocked LPS-induced pulmonary vascular leakage, alleviated acute lung injury, and improved survival in a mouse model of lethal LPS-induced endotoxemia. We found CitH3 in the bloodstream 30 min after intraperitoneal injection of LPS (35 mg/kg) into mice. Additionally, CitH3 production was induced in cultured neutrophils exposed to LPS, and NETs derived from these LPS-treated neutrophils increased the permeability of endothelial cells. However, YW3-56 reduced CitH3 production and NET formation by neutrophils following LPS exposure. Moreover, treatment with YW3-56 decreased the levels of circulating CitH3 and abolished neutrophil activation and NET formation in the lungs of mice with endotoxemia. These data suggest a novel mechanism by which PAD-NET-CitH3 can play a pivotal role in pulmonary vascular dysfunction and the pathogenesis of lethal endotoxemia.
Sigma-Aldrich
Lipopolysaccharides from Salmonella typhosa, purified by phenol extraction
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution