Merck
  • Probing the importance of the hemilabile site of bis(phosphine) monoxide ligands in the copper-catalyzed addition of diethylzinc to N-phosphinoylimines: discovery of new effective chiral ligands.
Probing the importance of the hemilabile site of bis(phosphine) monoxide ligands in the copper-catalyzed addition of diethylzinc to N-phosphinoylimines: discovery of new effective chiral ligands.
The Journal of organic chemistry (2008-07-22)
Isabelle Bonnaventure, André B Charette
ABSTRACT
The hemilabile ligand Me-DuPHOS(O) 2 has proven to be a successful ligand for the copper-catalyzed addition of diethylzinc to N-phosphinoylimines. The corresponding alpha-chiral amines were obtained in high yields (80-98%) and enantiomeric ratios (19.0:1 to 99.0:1 er). Furthermore, this Cu* 2 catalytic system has been shown to be effective in the addition of diethylzinc to nitroalkenes and in the reduction of beta,beta-disubstituted vinyl phenyl sulfones. This paper describes a general structure/selectivity study in which the three ligand subunits (chiral phospholane-linker-labile coordinating group (Z)) are systematically modified and tested in the copper-catalyzed addition of diethylzinc to the N-phosphinoylimine 1 derived from benzaldehyde. This study led to the discovery of a new class of effective chiral ligands that combine a chiral phospholane unit and an achiral phosphine oxide.
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc, packaged for use in deposition systems