Merck
  • Human Cord Blood Serum-Derived APP α-Secretase Cleavage Activity is Mediated by C1 Complement.
Human Cord Blood Serum-Derived APP α-Secretase Cleavage Activity is Mediated by C1 Complement.
Cell transplantation (2018-06-07)
Ahsan Habib, Darrell Sawmiller, Huayan Hou, Manasa Kanithi, Jun Tian, Jin Zeng, Dan Zi, Zhi-Xu He, Paul R Sanberg, Jun Tan
ABSTRACT
Alzheimer's Disease (AD) is the leading cause of dementia in the elderly. In healthy individuals, amyloid precursor protein (APP) is cleaved by α-secretase, generating soluble α-amyloid precursor protein (sAPPα), which contributes neuroprotective functions in the neuronal environment. In contrast, in the neurodegenerative environment of AD patients, amyloid-β-peptide (Aβ) of either 40 or 42 residues are generated by increased activity of β- and γ-secretase. These proteins amalgamate in specific regions of the brain, which disrupts neuronal functions and leads to cognitive impairment. Human umbilical cord blood cells (HUCBC) have proven useful as potential immunomodulatory therapies in various models of neurodegenerative diseases, including AD. Our most recent work studied the impact of umbilical cord blood serum (CBS) on modulation of sAPPα production. Heat-sensitive CBS significantly promoted sAPPα production, indicating that heat-sensitive factor(s) play(s) a role in this process. Liquid chromatography with tandem mass spectrometry (LC-MS/MS) analysis was used to determine the molecular source of α-secretase in purified CBS and aged blood serum (AgBS) fraction. Of the proteins identified, the subunits of C1 complex (C1q, C1r, and C1s) and alpha-2-macroglobulin showed significantly greater levels in purified α-CBS fraction (α-CBSF) compared with the AgBS fraction (AgBSF). Specifically, C1 markedly increased sAPPα and alpha-carboxyl-terminal fragment (α-CTF) production in a dose-dependent fashion, whereas C1q alone only minimally increased and C3 did not increase sAPPα production in the absence of sera. Furthermore, C1q markedly increased sAPPα and α-CTF, while decreasing Aβ, in CHO/APPwt cells cultured in the presence of whole sera. These results confirm our initial assumption that APP α-secretase activity in human blood serum is mediated by complement C1, opening a potential therapeutic modality for the future of AD.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Human APP / Amyloid Beta A4 Protein ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
C1 Inhibitor,
Human Recombinant
Sigma-Aldrich
Complement C3-Depleted Serum, Human
Sigma-Aldrich
Complement C1, Human, Complement C1, Human Native, is the first component of the classical complement pathway. It is a calcium-dependent complex of C1q, C1r, and C1s subcomponents.