Merck
  • Interleukin-21 and the IL-21 receptor: novel effectors of NK and T cell responses.
Interleukin-21 and the IL-21 receptor: novel effectors of NK and T cell responses.
Journal of leukocyte biology (2002-11-14)
Julia Parrish-Novak, Donald C Foster, Richard D Holly, Christopher H Clegg
ABSTRACT
Interleukin (IL)-21 was recently discovered using a functional cloning approach based on expression of its receptor. It is similar in domain organization and primary sequence to IL-2 and IL-15. Like these cytokines, IL-21 uses the common gamma chain of the IL-2/15 receptor, which forms a heterodimeric receptor complex with IL-21R. IL-21 is produced by activated T cells, and it influences proliferation of T and B cells and cytolytic activity of natural killer cells. The elucidation of the unique biological effects of IL-21 represents an intense area of interest in current cytokine biology.
Sigma-Aldrich
IL-21 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-21 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture