Merck
EGFR Induces E2F1-Mediated Corticotroph Tumorigenesis.
Journal of the Endocrine Society (2017-12-22)
Takako Araki, Xiaohai Liu, Hiraku Kameda, Yukiko Tone, Hidenori Fukuoka, Masahide Tone, Shlomo Melmed
ABSTRACT
The epidermal growth factor receptor (EGFR), expressed in adrenocorticotrophic hormone (ACTH)-secreting pituitary adenomas causing Cushing disease, regulates ACTH production and corticotroph proliferation. To elucidate the utility of EGFR as a therapeutic target for Cushing disease, we generated transgenic (Tg) mice with corticotroph-specific human EGFR expression (corti-EGFR-Tg) using a newly constructed corticotroph-specific promoter. Pituitary-specific EGFR expression was observed by 2.5 months, and aggressive ACTH-secreting pituitary adenomas with features of Crooke's cells developed by 8 months with 65% penetrance observed. Features consistent with the Cushing phenotype included elevated plasma ACTH and corticosterone levels, increased body weight, glucose intolerance, and enlarged adrenal cortex. Gefitinib, an EGFR tyrosine kinase inhibitor, suppressed tumor POMC expression and downstream EGFR tumor signaling, and ACTH and corticosterone levels were attenuated by 80% and 78%, respectively. Both E2F1 and phosphorylated Ser-337 E2F1 were increased in corti-EGFR-Tg mice and also colocalized with human POMC (hPOMC) in human pituitary corticotroph tumor samples. EGFR inhibition reversed E2F1 activity in vivo, whereas E2F1 inhibition suppressed POMC and ACTH in cultured human pituitary tumor cells. The corti-EGFR-Tg phenotype recapitulates ACTH-secreting pituitary adenomas and Cushing disease, validating the relevance of EGFR to corticotroph tumorigenesis. E2F1 is identified as a promising corticotroph-specific target for ACTH-dependent Cushing disease.
Sigma-Aldrich
Anti-Dopamine D2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid