Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc sulfate monohydrate
Linear Formula:
ZnSO
4
· H
2
O
CAS Number:
7446-19-7
Molecular Weight:
179.47
EC Number:
231-793-3
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
307491
Zinc sulfate monohydrate,
≥99.9% trace metals basis
Expand
View Pricing
96495
Zinc sulfate monohydrate,
purum p.a., ≥99.0% (KT)
Expand
View Pricing